LIF444784076 Life Chemicals Inc.

3-oxopiperazine-1-sulfonamide

  • MDL number MFCD11205214
  • Cas Number : 878386-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart