LIF444784079 Life Chemicals Inc.

6-(piperazin-1-yl)-N-(p-tolyl)pyridazin-3-amine

  • MDL number MFCD09881150
  • Cas Number : 1023812-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart