LIF444784088 Life Chemicals Inc.

2-[4-(propan-2-yl)piperazine-1-carbonyl]benzoic acid

  • MDL number MFCD05862468
  • Cas Number : 890092-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart