LIF444784089 Life Chemicals Inc.

2-(4-cyclopentylpiperazine-1-carbonyl)benzoic acid

  • MDL number MFCD05862478
  • Cas Number : 890092-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart