LIF444784091 Life Chemicals Inc.

6-methyl-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione

  • MDL number MFCD06809783
  • Cas Number : 4692-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart