LIF444784102 Life Chemicals Inc.

4-methyl-5-(1H-pyrrol-1-yl)nicotinic acid

  • MDL number MFCD28165970
  • Cas Number : 1820609-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart