LIF444784107 Life Chemicals Inc.

8-methoxy-6-methyl-1,2,3,4-tetrahydro-1,7-naphthyridine

  • MDL number MFCD28388527
  • Cas Number : 1517043-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart