LIF444784109 Life Chemicals Inc.

5-bromo-8-chloro-6-methyl-1,7-naphthyridine

  • MDL number MFCD27986222
  • Cas Number : 1439441-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart