LIF444784110 Life Chemicals Inc.

6,8-dimethyl-2H-benzo[d][1,3]oxazine-2,4(1H)-dione

  • MDL number MFCD03839912
  • Cas Number : 56934-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart