LIF444784112 Life Chemicals Inc.

2,6,8-trimethylquinoline-4-carboxylic acid

  • MDL number MFCD02741572
  • Cas Number : 93189-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart