LIF444784116 Life Chemicals Inc.

4-methoxy-3-(methoxymethyl)benzaldehyde

  • MDL number MFCD01822195
  • Cas Number : 76646-41-8
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart