LIF444784118 Life Chemicals Inc.

3-(aminomethyl)-4-methoxybenzaldehyde hydrochloride

  • MDL number MFCD01902022
  • Cas Number : 1820735-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart