LIF444784122 Life Chemicals Inc.

3-((dimethylamino)methyl)-4-methoxybenzaldehyde hydrochloride

  • MDL number MFCD07357556
  • Cas Number : 1177308-09-6
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart