LIF444784125 Life Chemicals Inc.

(2-aminophenyl)(p-tolyl)methanone

  • MDL number MFCD00007716
  • Cas Number : 36192-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart