LIF444784133 Life Chemicals Inc.

[1-(propan-2-yl)-1H-imidazol-2-yl]methanol

  • MDL number MFCD11108594
  • Cas Number : 135205-82-2
  • Molecular Weight : 140.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart