LIF444784134 Life Chemicals Inc.

(1-propyl-1H-imidazol-2-yl)methanol

  • MDL number MFCD09881599
  • Cas Number : 143886-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart