LIF444784139 Life Chemicals Inc.

2-(5-fluoro-1H-benzo[d]imidazol-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD09029275
  • Cas Number : 1187582-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart