LIF444784141 Life Chemicals Inc.

(2-amino-4-chlorophenyl)(phenyl)methanol

  • MDL number MFCD21110015
  • Cas Number : 25648-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart