LIF444784146 Life Chemicals Inc.

2,4,6-trimethylquinoline

  • MDL number MFCD00209450
  • Cas Number : 2243-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart