LIF444784156 Life Chemicals Inc.

4-methoxy-2-(methylsulfonyl)benzo[d]thiazole

  • MDL number MFCD22587563
  • Cas Number : 1334368-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart