LIF444784160 Life Chemicals Inc.

4,6-difluoro-N-((tetrahydrofuran-2-yl)methyl)benzo[d]thiazol-2-amine

  • MDL number MFCD16631662
  • Cas Number : 1219914-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart