LIF444784173 Life Chemicals Inc.

N1-(4-methoxy-7-methylbenzo[d]thiazol-2-yl)-N2,N2-dimethylethane-1,2-diamine

  • MDL number MFCD22587579
  • Cas Number : 1251544-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart