LIF444784174 Life Chemicals Inc.

N1-(5-chloro-4-methylbenzo[d]thiazol-2-yl)-N3,N3-dimethylpropane-1,3-diamine

  • MDL number MFCD22587580
  • Cas Number : 1251614-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart