LIF444784175 Life Chemicals Inc.

N1-(7-chloro-4-methylbenzo[d]thiazol-2-yl)-N3,N3-dimethylpropane-1,3-diamine

  • MDL number MFCD22587581
  • Cas Number : 1251614-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart