LIF444784179 Life Chemicals Inc.

6-ethoxy-N-(furan-2-ylmethyl)benzo[d]thiazol-2-amine

  • MDL number MFCD22587629
  • Cas Number : 1286732-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart