LIF444784180 Life Chemicals Inc.

3-ethyl-4,6-difluorobenzo[d]thiazol-2(3H)-imine

  • MDL number MFCD22587632
  • Cas Number : 1286728-37-7
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart