LIF444784181 Life Chemicals Inc.

5-phenyl-4-(p-tolyl)thiazol-2-amine

  • MDL number MFCD00553254
  • Cas Number : 75745-30-1
  • Molecular Weight : 266.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart