LIF444784182 Life Chemicals Inc.

2-amino-N,6-dimethyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxamide

  • MDL number MFCD11651567
  • Cas Number : 1099624-26-6
  • Molecular Weight : 225.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart