LIF444784184 Life Chemicals Inc.

5,6-dimethyl-N-(3-morpholinopropyl)benzo[d]thiazol-2-amine

  • MDL number MFCD22587637
  • Cas Number : 1286725-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart