LIF444784193 Life Chemicals Inc.

2-amino-3-(2-methoxy-2-oxoethyl)-6,7-dihydro-[1,4]dioxino[2',3':4,5]benzo[1,2-d]thiazol-3-ium bromide

  • MDL number MFCD22587661
  • Cas Number : 2197055-18-6
  • Molecular Weight : 361.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart