LIF444784209 Life Chemicals Inc.

ethyl 2-(4,6-difluoro-2-iminobenzo[d]thiazol-3(2H)-yl)acetate hydrobromide

  • MDL number MFCD22587722
  • Cas Number : 1351654-38-0
  • Molecular Weight : 353.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart