LIF444784210 Life Chemicals Inc.

N-(3-allyl-2-imino-2,3-dihydrobenzo[d]thiazol-6-yl)acetamide hydrobromide

  • MDL number MFCD22587724
  • Cas Number : 1351645-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart