LIF444784212 Life Chemicals Inc.

3-allylbenzo[d]thiazol-2(3H)-imine hydrobromide

  • MDL number MFCD22587727
  • Cas Number : 175920-97-5
  • Molecular Weight : 271.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart