LIF444784213 Life Chemicals Inc.

6-nitro-3-(prop-2-yn-1-yl)benzo[d]thiazol-2(3H)-imine hydrobromide

  • MDL number MFCD22587729
  • Cas Number : 1351597-37-9
  • Molecular Weight : 314.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart