LIF444784220 Life Chemicals Inc.

5-(7-ethoxybenzofuran-2-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD22587793
  • Cas Number : 1421485-28-0
  • Molecular Weight : 245.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart