LIF444784223 Life Chemicals Inc.

1-(6-methoxybenzo[d]thiazol-2-yl)piperidine-4-carboxylic acid

  • MDL number MFCD15730074
  • Cas Number : 1282321-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart