LIF444784225 Life Chemicals Inc.

7-chloro-3,4-dimethylbenzo[d]thiazol-2(3H)-imine

  • MDL number MFCD22587796
  • Cas Number : 782391-47-3
  • Molecular Weight : 212.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart