LIF444784241 Life Chemicals Inc.

ethyl 2-amino-1-benzothiophene-3-carboxylate

  • MDL number MFCD00102506
  • Cas Number : 7311-95-7
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart