LIF444784246 Life Chemicals Inc.

6-(1,3-dioxoisoindolin-2-yl)hexanoic acid

  • MDL number MFCD00233499
  • Cas Number : 4443-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart