LIF444784263 Life Chemicals Inc.

1-(5-hydroxy-2-methyl-1-phenyl-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD00561131
  • Cas Number : 5102-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart