LIF444784273 Life Chemicals Inc.

4-(4-nitrothiophen-2-yl)thiazol-2-amine

  • MDL number MFCD01681537
  • Cas Number : 58139-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart