LIF444784276 Life Chemicals Inc.

1-propyl-1,3-benzodiazole-2-thiol

  • MDL number MFCD01008568
  • Cas Number : 67624-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart