LIF444784279 Life Chemicals Inc.

N-(4-methoxybenzo[d]thiazol-2-yl)-7-(methylthio)benzo[1,2-d:4,3-d']bis(thiazole)-2-amine

  • MDL number MFCD05892625
  • Cas Number : 862974-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart