LIF444784281 Life Chemicals Inc.

N-benzyl-4-fluorobenzo[d]thiazol-2-amine

  • MDL number MFCD05892684
  • Cas Number : 862975-05-1
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart