LIF444784296 Life Chemicals Inc.

1-((9H-fluoren-9-yl)methyl)piperidine

  • MDL number MFCD22587875
  • Cas Number : 35661-58-6
  • Molecular Weight : 263.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart