LIF444784306 Life Chemicals Inc.

4-(aminomethyl)-2-fluorobenzonitrile

  • MDL number MFCD06658314
  • Cas Number : 368426-73-7
  • Molecular Weight : 150.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart