LIF444784312 Life Chemicals Inc.

3-(benzo[d][1,3]dioxol-5-yl)pyrrolidine

  • MDL number MFCD11613467
  • Cas Number : 1082806-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart