LIF444784313 Life Chemicals Inc.

2-(4-methoxyphenyl)morpholine

  • MDL number MFCD03840011
  • Cas Number : 83555-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart