LIF444784328 Life Chemicals Inc.

1-ethyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD04968670
  • Cas Number : 957514-21-5
  • Molecular Weight : 194.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart