LIF444784329 Life Chemicals Inc.

4-fluoro-3-(1H-tetrazol-5-yl)benzenesulfonyl chloride

  • MDL number MFCD09044808
  • Cas Number : 926222-53-9
  • Molecular Weight : 262.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart